Anti-EXD1 Mouse Monoclonal Antibody [clone: OTI1G1]

Toimittaja: OriGene
ORIGTA502102EA 581 EUR
ORIGTA502102
Anti-EXD1 Mouse Monoclonal Antibody [clone: OTI1G1]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: EXD1 (exonuclease 3'-5' domain containing 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR