Anti-HDAC6 Mouse Monoclonal Antibody [clone: OTI3E7]

Toimittaja: OriGene
ORIGTA502177EA 603 EUR
ORIGTA502177
Anti-HDAC6 Mouse Monoclonal Antibody [clone: OTI3E7]
Vasta-aineet
  • Recommended Dilutions: Western Blot: 1:500-1000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: HDAC6 (histone deacetylase 6)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR