Anti-CDCP1 Mouse Monoclonal Antibody [clone: OTI5B3]

Toimittaja: OriGene
ORIGTA502231EA 608 EUR
ORIGTA502231
Anti-CDCP1 Mouse Monoclonal Antibody [clone: OTI5B3]
Vasta-aineet
  • Recommended Dilutions: Western Blot: 1:500-1000; Flow Cytometry: 1:100

Type: Primary
Antigen: CDCP1 (CUB domain containing protein 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR