Anti-MMAA Mouse Monoclonal Antibody [clone: OTI3A3]

Toimittaja: OriGene
ORIGTA502278EA 586 EUR
ORIGTA502278
Anti-MMAA Mouse Monoclonal Antibody [clone: OTI3A3]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:200-500; Immunochemistry: 1:150

Type: Primary
Antigen: MMAA (methylmalonic aciduria (cobalamin deficiency) cblA type)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR