Anti-CTNNB1 Mouse Monoclonal Antibody [clone: OTI6E7]

Toimittaja: OriGene
ORIGTA502305EA 603 EUR
ORIGTA502305
Anti-CTNNB1 Mouse Monoclonal Antibody [clone: OTI6E7]
Vasta-aineet
  • Recommended Dilutions: Western Blot: 1:500-2000; Flow Cytometry: 1:100

Type: Primary
Antigen: CTNNB1 (catenin (cadherin-associated protein), beta 1, 88kDa)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR