Anti-DNAJB2 Mouse Monoclonal Antibody [clone: OTI5C11]

Toimittaja: OriGene
ORIGTA502329EA 603 EUR
ORIGTA502329
Anti-DNAJB2 Mouse Monoclonal Antibody [clone: OTI5C11]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: DNAJB2 (DnaJ (Hsp40) homolog, subfamily B, member 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR