Anti-CHCHD5 Mouse Monoclonal Antibody [clone: OTI4B3]

Toimittaja: OriGene
ORIGTA502350EA 603 EUR
ORIGTA502350
Anti-CHCHD5 Mouse Monoclonal Antibody [clone: OTI4B3]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500; Flow Cytometry: 1:100

Type: Primary
Antigen: CHCHD5 (coiled-coil-helix-coiled-coil-helix domain containing 5)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR