Anti-AQP1 Mouse monoclonal antibody [clone: OTI2D10]

Toimittaja: OriGene
ORIGTA502357EA 609 EUR
ORIGTA502357
Anti-AQP1 Mouse monoclonal antibody [clone: OTI2D10]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: AQP1 (aquaporin 1 (Colton blood group))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR