Anti-PRKCE Mouse Monoclonal Antibody [clone: OTI3F10]
Toimittaja: OriGene
ORIGTA502368EA 603 EUR
ORIGTA502368
Anti-PRKCE Mouse Monoclonal Antibody [clone: OTI3F10]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100
Type: Primary
Antigen: PRKCE (protein kinase C, epsilon)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human