Anti-EIF4E2 Mouse Monoclonal Antibody [clone: OTI4E7]

Toimittaja: OriGene
ORIGTA502431EA 603 EUR
ORIGTA502431
Anti-EIF4E2 Mouse Monoclonal Antibody [clone: OTI4E7]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: EIF4E2 (eukaryotic translation initiation factor 4E family member 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR