Anti-VASP Mouse Monoclonal Antibody [clone: OTI2G1]

Toimittaja: OriGene
ORIGTA502472EA 581 EUR
ORIGTA502472
Anti-VASP Mouse Monoclonal Antibody [clone: OTI2G1]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: VASP (vasodilator-stimulated phosphoprotein)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR