Anti-RASSF1 Mouse Monoclonal Antibody [clone: OTI2A9]

Toimittaja: OriGene
ORIGTA502481EA 603 EUR
ORIGTA502481
Anti-RASSF1 Mouse Monoclonal Antibody [clone: OTI2A9]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: RASSF1 (Ras association (RalGDS/AF-6) domain family member 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR