Anti-CAT Mouse Monoclonal Antibody [clone: OTI1B6]

Toimittaja: OriGene
ORIGTA502496EA 608 EUR
ORIGTA502496
Anti-CAT Mouse Monoclonal Antibody [clone: OTI1B6]
Vasta-aineet
  • Recommended Dilutions: Western Blot: 1:200-500; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: CAT (catalase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR