Anti-ANKRD53 Mouse Monoclonal Antibody [clone: OTI1A7]

Toimittaja: OriGene
ORIGTA502505EA 603 EUR
ORIGTA502505
Anti-ANKRD53 Mouse Monoclonal Antibody [clone: OTI1A7]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:1000-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: ANKRD53 (ankyrin repeat domain 53)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR