Anti-NECAB1 Mouse Monoclonal Antibody [clone: OTI1H5]

Toimittaja: OriGene
ORIGTA502520EA 603 EUR
ORIGTA502520
Anti-NECAB1 Mouse Monoclonal Antibody [clone: OTI1H5]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: NECAB1 (N-terminal EF-hand calcium binding protein 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR