Anti-HOXC11 Mouse Monoclonal Antibody [clone: OTI3E5]

Toimittaja: OriGene
ORIGTA502579EA 593 EUR
ORIGTA502579
Anti-HOXC11 Mouse Monoclonal Antibody [clone: OTI3E5]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: HOXC11 (homeobox C11)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR