Anti-ANKRD53 Mouse Monoclonal Antibody [clone: OTI2F3]

Toimittaja: OriGene
ORIGTA502598EA 603 EUR
ORIGTA502598
Anti-ANKRD53 Mouse Monoclonal Antibody [clone: OTI2F3]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: ANKRD53 (ankyrin repeat domain 53)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR