Anti-DSTN Mouse Monoclonal Antibody [clone: OTI1D5]

Toimittaja: OriGene
ORIGTA502653EA 603 EUR
ORIGTA502653
Anti-DSTN Mouse Monoclonal Antibody [clone: OTI1D5]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: DSTN (destrin (actin depolymerizing factor))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR