Anti-NDUFA7 Mouse Monoclonal Antibody [clone: OTI2D4]

Toimittaja: OriGene
ORIGTA502689EA 603 EUR
ORIGTA502689
Anti-NDUFA7 Mouse Monoclonal Antibody [clone: OTI2D4]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: NDUFA7 (NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 7, 14.5kDa)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR