Anti-MAB21L3 Mouse Monoclonal Antibody [clone: OTI1F6]

Toimittaja: OriGene
ORIGTA502716EA 586 EUR
ORIGTA502716
Anti-MAB21L3 Mouse Monoclonal Antibody [clone: OTI1F6]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: MAB21L3 (mab-21-like 3 (C. elegans))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR