Anti-TP53 Mouse Monoclonal Antibody [clone: OTI1F4]

Toimittaja: OriGene
ORIGTA502802EA 603 EUR
ORIGTA502802
Anti-TP53 Mouse Monoclonal Antibody [clone: OTI1F4]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: TP53 (tumor protein p53)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR