Anti-ASCC2 Mouse Monoclonal Antibody [clone: OTI1E8]

Toimittaja: OriGene
ORIGTA502806EA 608 EUR
ORIGTA502806
Anti-ASCC2 Mouse Monoclonal Antibody [clone: OTI1E8]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: ASCC2 (activating signal cointegrator 1 complex subunit 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR