Anti-FGFR2 Mouse Monoclonal Antibody [clone: OTI2D1]

Toimittaja: OriGene
ORIGTA502843EA 603 EUR
ORIGTA502843
Anti-FGFR2 Mouse Monoclonal Antibody [clone: OTI2D1]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: FGFR2 (fibroblast growth factor receptor 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR