Anti-CDK2 Mouse Monoclonal Antibody [clone: OTI2D9]

Toimittaja: OriGene
ORIGTA502935EA 608 EUR
ORIGTA502935
Anti-CDK2 Mouse Monoclonal Antibody [clone: OTI2D9]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: CDK2 (cyclin-dependent kinase 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR