Anti-SOCS3 Mouse Monoclonal Antibody [clone: OTI11C5]

Toimittaja: OriGene
ORIGTA502959EA 603 EUR
ORIGTA502959
Anti-SOCS3 Mouse Monoclonal Antibody [clone: OTI11C5]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: SOCS3 (suppressor of cytokine signaling 3)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR