Anti-SNAP25 Mouse Monoclonal Antibody [clone: OTI1G2]
Toimittaja: OriGene
ORIGTA502967EA 603 EUR
ORIGTA502967
Anti-SNAP25 Mouse Monoclonal Antibody [clone: OTI1G2]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100
Type: Primary
Antigen: SNAP25 (synaptosomal-associated protein, 25kDa)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human