Anti-ITFG2 Mouse Monoclonal Antibody [clone: OTI3A5]

Toimittaja: OriGene
ORIGTA503064EA 581 EUR
ORIGTA503064
Anti-ITFG2 Mouse Monoclonal Antibody [clone: OTI3A5]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500

Type: Primary
Antigen: ITFG2 (integrin alpha FG-GAP repeat containing 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR