Anti-PHF21B Mouse Monoclonal Antibody [clone: OTI3G9]

Toimittaja: OriGene
ORIGTA503072EA 603 EUR
ORIGTA503072
Anti-PHF21B Mouse Monoclonal Antibody [clone: OTI3G9]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500; Flow Cytometry: 1:100

Type: Primary
Antigen: PHF21B (PHD finger protein 21B)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR