Anti-ILVBL Mouse Monoclonal Antibody [clone: OTI8B12]

Toimittaja: OriGene
ORIGTA503079EA 603 EUR
ORIGTA503079
Anti-ILVBL Mouse Monoclonal Antibody [clone: OTI8B12]
Vasta-aineet
  • Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: ILVBL (ilvB (bacterial acetolactate synthase)-like)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR