Anti-NUDT9 Mouse Monoclonal Antibody [clone: OTI7F12]

Toimittaja: OriGene
ORIGTA503235EA 593 EUR
ORIGTA503235
Anti-NUDT9 Mouse Monoclonal Antibody [clone: OTI7F12]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: NUDT9 (nudix (nucleoside diphosphate linked moiety X)-type motif 9)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR