Anti-PDE1B Mouse Monoclonal Antibody [clone: OTI6E4]

Toimittaja: OriGene
ORIGTA503319EA 616 EUR
ORIGTA503319
Anti-PDE1B Mouse Monoclonal Antibody [clone: OTI6E4]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: PDE1B (phosphodiesterase 1B, calmodulin-dependent)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR