Anti-HMOX2 Mouse Monoclonal Antibody [clone: OTI4E10]

Toimittaja: OriGene
ORIGTA503821EA 593 EUR
ORIGTA503821
Anti-HMOX2 Mouse Monoclonal Antibody [clone: OTI4E10]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: HMOX2 (heme oxygenase (decycling) 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR