Anti-EXOSC7 Mouse Monoclonal Antibody [clone: OTI1E8]

Toimittaja: OriGene
ORIGTA504228EA 616 EUR
ORIGTA504228
Anti-EXOSC7 Mouse Monoclonal Antibody [clone: OTI1E8]
Vasta-aineet
  • Recommended Dilutions: Western Blot: 1:4000

Type: Primary
Antigen: EXOSC7 (exosome component 7)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR