Anti-SERPINF1 Mouse Monoclonal Antibody [clone: OTI1F7]

Toimittaja: OriGene
ORIGTA504434EA 621 EUR
ORIGTA504434
Anti-SERPINF1 Mouse Monoclonal Antibody [clone: OTI1F7]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: SERPINF1 (serpin peptidase inhibitor, clade F (alpha-2 antiplasmin, pigment epithelium derived factor), member 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR