Anti-CPO Mouse Monoclonal Antibody [clone: OTI4A9]

Toimittaja: OriGene
ORIGTA504593EA 616 EUR
ORIGTA504593
Anti-CPO Mouse Monoclonal Antibody [clone: OTI4A9]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: CPO (carboxypeptidase O)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR