Anti-FOLH1 Mouse monoclonal antibody [clone: OTI3H5]

Toimittaja: OriGene
ORIGTA504752EA 574 EUR
ORIGTA504752
Anti-FOLH1 Mouse monoclonal antibody [clone: OTI3H5]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-1000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: FOLH1 (folate hydrolase (prostate-specific membrane antigen) 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR