Anti-CNN2 Rabbit Polyclonal Antibody

Toimittaja: Abnova
ABNOH00001265-D01PEA 380 EUR
ABNOH00001265-D01P
Anti-CNN2 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: CNN2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR