Anti-SPP1 Mouse Monoclonal Antibody [clone: 3G6]

Toimittaja: Abnova
ABNOH00006696-M17EA 380 EUR
ABNOH00006696-M17
Anti-SPP1 Mouse Monoclonal Antibody [clone: 3G6]
Vasta-aineet
Type: Primary
Antigen: SPP1
Clonality: Monoclonal
Clone: 3G6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR