Anti-TARDBP Mouse Monoclonal Antibody [clone: 2E2-D3]

Toimittaja: Abnova
ABNOH00023435-M01EA 394 EUR
ABNOH00023435-M01
Anti-TARDBP Mouse Monoclonal Antibody [clone: 2E2-D3]
Vasta-aineet
Type: Primary
Antigen: TARDBP
Clonality: Monoclonal
Clone: 2E2-D3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR