Anti-TPRKB Mouse Monoclonal Antibody [clone: 3F6]

Toimittaja: Abnova
ABNOH00051002-M01EA 380 EUR
ABNOH00051002-M01
Anti-TPRKB Mouse Monoclonal Antibody [clone: 3F6]
Vasta-aineet
Type: Primary
Antigen: TPRKB
Clonality: Monoclonal
Clone: 3F6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR