Anti-PVRIG Mouse Polyclonal Antibody

Toimittaja: Abnova
ABNOH00079037-B02PEA 394 EUR
ABNOH00079037-B02P
Anti-PVRIG Mouse Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: PVRIG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR