Anti-GPT2 Mouse Polyclonal Antibody

Toimittaja: Abnova
ABNOH00084706-B01PEA 394 EUR
ABNOH00084706-B01P
Anti-GPT2 Mouse Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: GPT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR