Anti-APP Rabbit polyclonal antibody

Toimittaja: Biorbyt
BIRBORB4527-100EA 334 EUR
BIRBORB4527-100
Anti-APP Rabbit polyclonal antibody
Vasta-aineet
  • ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: APP (amyloid beta (A4) precursor protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR