Anti-SGMS1 Rabbit Polyclonal Antibody

Toimittaja: Biorbyt
BIRBORB4556-200EA 0 EUR
BIRBORB4556-200 BIRBORB4556-100
Anti-SGMS1 Rabbit Polyclonal Antibody
Vasta-aineet
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: SGMS1 (sphingomyelin synthase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR