Anti-ATM Rabbit polyclonal antibody

Toimittaja: Biorbyt
BIRBORB4605-100EA 0 EUR
BIRBORB4605-100
Anti-ATM Rabbit polyclonal antibody
Vasta-aineet
Immunochemistry: 1:50-1:100

Type: Primary
Antigen: ATM (ataxia telangiectasia mutated)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR