Anti-BLNK Rabbit Polyclonal Antibody
Toimittaja: Biorbyt
BIRBORB4693-100EA 334 EUR
BIRBORB4693-100
Anti-BLNK Rabbit Polyclonal Antibody
Vasta-aineet
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: BLNK (B-cell linker)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human