Anti-NPTX1 Rabbit Polyclonal Antibody
Toimittaja: Biorbyt
BIRBORB4810-200EA 0 EUR
BIRBORB4810-200 BIRBORB4810-100
Anti-NPTX1 Rabbit Polyclonal Antibody
Vasta-aineet
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: NPTX1 (neuronal pentraxin I)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat