Anti-CCT8L2 Rabbit Polyclonal Antibody (FITC (Fluorescein))

Toimittaja: Biorbyt
BIRBORB4831-100EA 0 EUR
BIRBORB4831-100
Anti-CCT8L2 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Vasta-aineet
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)

Type: Primary
Antigen: CCT8L2 (chaperonin containing TCP1, subunit 8 (theta)-like 2)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR