Anti-ITGA2B Rabbit Polyclonal Antibody

Toimittaja: Biorbyt
BIRBORB4832-100EA 312 EUR
BIRBORB4832-100 BIRBORB4832-200
Anti-ITGA2B Rabbit Polyclonal Antibody
Vasta-aineet
Western Blot: 500-1500; Immunochemistry: 100 -1000; Immunofluorescence: 1:200

Type: Primary
Antigen: ITGA2B (integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigen CD41))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR