Anti-RBX1 Rabbit Polyclonal Antibody (FITC (Fluorescein))

Toimittaja: Biorbyt
BIRBORB5247-100EA 0 EUR
BIRBORB5247-100
Anti-RBX1 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Vasta-aineet
Type: Primary
Antigen: RBX1 (ring-box 1, E3 ubiquitin protein ligase)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR